
(+)-Menthofuran

Synonym(s):
(6R)-4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran, (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing