Skip to Content
Merck
Discovery® HS C18 Prep Guard Cartridge 10 μm particle size, L × I.D. 1 cm × 21.2 mm

Discovery® HS C18 (10 µm) HPLC Columns

Discovery® HS C18 Prep Guard Cartridge 10 μm particle size, L × I.D. 1 cm × 21.2 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2