
Dipyridamole Related Compound C

Synonym(s):
2,2′-[(6-Chloro-4,8-di-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl)imino]bisethanol, 2,2′-{[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]azanediyl}diethanol, 2-Diethanolamino-6-chloro-4,8-dipiperidinopyrimido[5,4-d]pyrimidine
Empirical Formula (Hill Notation):
C20H30ClN7O2
CAS Number:
Molecular Weight:
435.95
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2