
Digalacturonic acid

Synonym(s):
α-D-GalA-(1→4)-D-GalA
Empirical Formula (Hill Notation):
C12H18O13
CAS Number:
Molecular Weight:
370.26
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing