Skip to Content
Merck
Diethylene glycol dibutyl ether ≥99%

Diethylene glycol dibutyl ether

Diethylene glycol dibutyl ether ≥99%
Synonym(s):
2-Butoxyethyl ether, Bis(2-butoxyethyl) ether, Dibutyldiglycol
Linear Formula:
(CH3CH2CH2CH2OCH2CH2)2O
CAS Number:
Molecular Weight:
218.33
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing