Skip to Content
Merck
Dicyclopentadiene contains BHT as stabilizer

Dicyclopentadiene

Dicyclopentadiene contains BHT as stabilizer
Synonym(s):
4,7-Methano-3a,4,7,7a-tetrahydroindene, Cyclopentadiene dimer, DCPD
Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing