Skip to Content
BN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dibutyl sulfide
Synonym(s):
1,1′-Thiobisbutane, Butyl sulfide, NSC 8460
Linear Formula:
CH
3
(CH
2
)
3
S(CH
2
)
3
CH
3
CAS Number:
544-40-1
Molecular Weight:
146.29
EC Number:
208-870-5
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
certified reference material (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
51491
Dibutyl sulfide,
≥98.0% (GC)
Expand
View Pricing
B101796
Dibutyl sulfide,
96%
Expand
View Pricing
76693
Dibutyl sulfide,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing