
Dibenzocycloheptene

Synonym(s):
5H-Dibenzo[a,d]cycloheptene
Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing