Skip to Content
Merck
Cyanuric acid analytical standard

Cyanuric acid

Cyanuric acid analytical standard
Synonym(s):
1,3,5-Triazine-2,4,6-triol, 2,4,6-Trihydroxy-1,3,5-triazine, Cyanuric acid
Linear Formula:
C3N3(OH)3
CAS Number:
Molecular Weight:
129.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing