
Crotaline

Synonym(s):
Monocrotaline
Empirical Formula (Hill Notation):
C16H23NO6
CAS Number:
Molecular Weight:
325.36
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing