![Chiralyst P294 Umicore](/deepweb/assets/sigmaaldrich/product/structures/195/798/8ac03793-c7a7-43e8-918f-5345e4fb17e9/640/8ac03793-c7a7-43e8-918f-5345e4fb17e9.png)
Chiralyst P294
![Chiralyst P294 Umicore](/deepweb/assets/sigmaaldrich/product/structures/195/798/8ac03793-c7a7-43e8-918f-5345e4fb17e9/640/8ac03793-c7a7-43e8-918f-5345e4fb17e9.png)
Synonym(s):
(Acetylacetonato)(norbornadiene)rhodium(I), (Acetylacetonato)(bicyclo[2.2.1]hepta-2,5-diene)rhodium(I)
Empirical Formula (Hill Notation):
C12H15O2Rh
CAS Number:
Molecular Weight:
294.15
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing