
Capecitabine Related Compound B

Synonym(s):
5-Fluoro-5′-deoxyuridine, 5′dFUrd, Doxifluridine
Empirical Formula (Hill Notation):
C9H11FN2O5
CAS Number:
Molecular Weight:
246.19
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing