Skip to Content
Merck
β-Sitosterol primary reference standard

β-Sitosterol

β-Sitosterol primary reference standard
Synonym(s):
α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique