![Bovine Serum Albumin solution 30%±2% in 0.85% sodium chloride, aseptically filled](/deepweb/assets/sigmaaldrich/product/images/370/212/c5d44dfc-c4b3-46fd-b6b1-345f632bdabd/640/c5d44dfc-c4b3-46fd-b6b1-345f632bdabd.jpg)
Bovine Serum Albumin solution
![Bovine Serum Albumin solution 30%±2% in 0.85% sodium chloride, aseptically filled](/deepweb/assets/sigmaaldrich/product/images/370/212/c5d44dfc-c4b3-46fd-b6b1-345f632bdabd/640/c5d44dfc-c4b3-46fd-b6b1-345f632bdabd.jpg)
Synonym(s):
Albumin solution from bovine serum, BSA solution
CAS Number:
Products
Product Category
Brand
Biological Source
Feature
Physical Form
Product Line
Quality Segment
Geographic Origin
Sterilization Method
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing