
Boc-Gly-OH

Synonym(s):
N-(tert-Butoxycarbonyl)glycine, Boc-glycine, Boc-Gly-OH
Linear Formula:
(CH3)3COCONHCH2COOH
CAS Number:
Molecular Weight:
175.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing