
Bis(pinacolato)diboron

Synonym(s):
4,4,4′,4′,5,5,5′,5′-Octamethyl-2,2′-bi-1,3,2-dioxaborolane
Empirical Formula (Hill Notation):
C12H24B2O4
CAS Number:
Molecular Weight:
253.94
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing