
Betaxolol impurity A

Synonym(s):
(1-(4-Ethylphenoxy)-3-(isopropylamino)propan-2-ol)
Empirical Formula (Hill Notation):
C14H23NO2
CAS Number:
Molecular Weight:
237.34
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing