Skip to Content
Merck
β-Cyclodextrin Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)

β-Cyclodextrin

β-Cyclodextrin Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)
Synonym(s):
Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin
Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Sterility
Technique