
Bacoside A3

Synonym(s):
(3β,16β,23R)-16,23:16,30-Diepoxy-20-hydroxydammar-24-en-3-yl O-α-L-arabinofuranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside, Jujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside]
Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3