Skip to Content
Merck
Atropine solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Atropine solution

Atropine solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Atropine solution
Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique