![Ascorbic Acid Impurity F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/344/379/38f8fd8b-5c82-43ff-a497-f7bdb8db8cb2/640/38f8fd8b-5c82-43ff-a497-f7bdb8db8cb2.png)
Ascorbic Acid Impurity F
![Ascorbic Acid Impurity F Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/344/379/38f8fd8b-5c82-43ff-a497-f7bdb8db8cb2/640/38f8fd8b-5c82-43ff-a497-f7bdb8db8cb2.png)
Synonym(s):
D-(−)-Isoascorbic acid, D-erythro-Hex-2-enoic acid γ-lactone, D-Araboascorbic acid
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing