
ANTI-FLAG® High Sensitivity, M2 coated 96-well plates

Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk
Products
Product Category
Brand
Clonality
Isotype
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing