
Ammonium lauryl sulfate solution

Synonym(s):
Ammonium dodecyl sulfate, Dodecyl sulfate ammonium salt
Linear Formula:
CH3(CH2)11OSO3NH4
CAS Number:
Molecular Weight:
283.43
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing