Skip to Content
Merck
9-Methylanthracene 98%

9-Methylanthracene

9-Methylanthracene 98%
Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing