
7-Methoxycoumarin-4-acetic acid

Synonym(s):
7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid
Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing