
4-Nitroso-hydrochlorothiazide

Synonym(s):
4-Nitroso-HCT, 6-Chloro-4-nitroso-1,1-dioxo-2,3-dihydro-1λ6,2,4-benzothiadiazine-7-sulfonamide
CAS Number:
Products
Brand
Manufacturer
Product Line
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing