Skip to Content
Merck
4-Hydroxy-4-methyl-2-pentanone analytical standard

4-Hydroxy-4-methyl-2-pentanone

4-Hydroxy-4-methyl-2-pentanone analytical standard
Synonym(s):
Diacetone alcohol
Linear Formula:
(CH3)2C(OH)CH2COCH3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing