Skip to Content
Merck
4-Formyl-1-methylpyridinium benzenesulfonate ≥95.0%

4-Formyl-1-methylpyridinium benzenesulfonate

4-Formyl-1-methylpyridinium benzenesulfonate ≥95.0%
Empirical Formula (Hill Notation):
C13H13NO4S
CAS Number:
Molecular Weight:
279.31

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing