4-Chlorobenzophenone
Synonym(s):
(4-Chlorophenyl)phenylmethanone
Linear Formula:
ClC6H4COC6H5
CAS Number:
Molecular Weight:
216.66
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing