Skip to Content
Merck
4-(4-Nitrophenylazo)resorcinol Dye content 90 %

4-(4-Nitrophenylazo)resorcinol

4-(4-Nitrophenylazo)resorcinol Dye content 90 %
Synonym(s):
Azoviolet, Magneson I
Linear Formula:
O2NC6H4N=NC6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
259.22
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing