3,3-Bis(chloromethyl)oxetane
Empirical Formula (Hill Notation):
C5H8Cl2O
CAS Number:
Molecular Weight:
155.02
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing