Skip to Content
Merck
(2-Methylpropyl)boronic acid ≥95.0%

(2-Methylpropyl)boronic acid

(2-Methylpropyl)boronic acid ≥95.0%
Synonym(s):
Isobutaneboronic acid
Linear Formula:
(CH3)2CHCH2B(OH)2
CAS Number:
Molecular Weight:
101.94

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing