Skip to Content
Merck
2-Chlorobenzaldehyde 99%

2-Chlorobenzaldehyde

2-Chlorobenzaldehyde 99%
Linear Formula:
ClC6H4CHO
CAS Number:
Molecular Weight:
140.57
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing