
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol

Empirical Formula (Hill Notation):
C22H29N3O
CAS Number:
Molecular Weight:
351.49
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing