Skip to Content
Merck
2,2,2-Trichloroethanol ReagentPlus®, ≥99%

2,2,2-Trichloroethanol

2,2,2-Trichloroethanol ReagentPlus®, ≥99%
Synonym(s):
Trichloroethyl alcohol
Linear Formula:
Cl3CCH2OH
CAS Number:
Molecular Weight:
149.40
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing