
1(S),9(R)-(−)-Bicuculline methchloride

Linear Formula:
C21H20NO6Cl
CAS Number:
Molecular Weight:
417.84
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing