
1-Deoxynojirimycin hydrochloride

Synonym(s):
1,5-Dideoxy-1,5-imino-D-sorbitol hydrochloride
Empirical Formula (Hill Notation):
C6H13NO4 · HCl
CAS Number:
Molecular Weight:
199.63
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing