Skip to Content
Merck
1-Bromodecane 98%

1-Bromodecane

1-Bromodecane 98%
Synonym(s):
Decyl bromide
Linear Formula:
CH3(CH2)9Br
CAS Number:
Molecular Weight:
221.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing