Skip to Content
Merck
1,4,7-Triazacyclononane 95%

1,4,7-Triazacyclononane

1,4,7-Triazacyclononane 95%
Synonym(s):
Octahydro-1H-1,4,7-triazonine, Triethylenetriamine
Empirical Formula (Hill Notation):
C6H15N3
CAS Number:
Molecular Weight:
129.20

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing