Skip to Content
Merck
1,3-Dimethyl-2-imidazolidinone reagent grade

1,3-Dimethyl-2-imidazolidinone

1,3-Dimethyl-2-imidazolidinone reagent grade
Synonym(s):
N,N′-Dimethylethyleneurea, DMEU, DMI
Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade