Skip to Content
Merck
1,2-Benzenedimethanol 97%

1,2-Benzenedimethanol

1,2-Benzenedimethanol 97%
Synonym(s):
1,2-Bis(hydroxymethyl)benzene, o-Xylene-α,α′-diol, Phthalyl alcohol
Linear Formula:
C6H4(CH2OH)2
CAS Number:
Molecular Weight:
138.16
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing