Skip to Content
Merck
All Photos(2)

Key Documents

Z732664

IKA® ULTRA-TURRAX® disperser tools, stainless steel

dispersing element for T-18 basic disperser, 10 mm, 1/cs

Synonym(s):

disperser blades, tissue grinders, tissue homogenizers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
NACRES:
NB.22

material

stainless steel

feature

stainless steel rotor/stator

packaging

case of 1
pack of 1

manufacturer/tradename

IKA L004639

diam.

10 mm

Looking for similar products? Visit Product Comparison Guide

General description

IKA® S 18 N - 10 G Dispersing tool for use with T-18 ULATRA-TURRAX® dispersing instrument.

Features and Benefits

  • suitable for use with solvents
  • sterilization methods: all methods

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service