Key Documents
Z360295
Carboy spigots
Tefzel®
Synonym(s):
plastic spigot, plastic spigots
About This Item
UNSPSC Code:
41121820
NACRES:
NB.35
material
Tefzel®
manufacturer/tradename
Nalgene®
Related Categories
General description
Spigots fit all Nalgene carboys. Both are autoclavable when installed on autoclavable carboys. Thread is 11/8 in. - 12 straight female threads. Spout accepts 5/8 in. I.D. tubing.
Tefzel® ETPE for use with solvents and other chemicals where a PP spigot is unsatisfactory.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Visit the Document Library
Customers Also Viewed
Slide 1 of 1