Skip to Content
Merck
All Photos(1)

Key Documents

SML1231

Sigma-Aldrich

c-di-AMP sodium salt

≥98% (HPLC), powder, bacterial second messenger

Synonym(s):

3′,5′-Cyclic diadenylic acid sodium salt, Adenylyl-(3′5′)-3′-adenylic acid, cyclic nucleotide sodium salt, Cyclic di-3′,5′-adenylate sodium salt, Cyclic diadenylate monophosphate sodium salt, Cyclic diadenylate sodium salt, Cyclic-di-AMP sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N10O12P2 · xNa+
CAS Number:
Molecular Weight:
658.41 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

c-di-AMP sodium salt, ≥98% (HPLC)

description

contains 1μmol of powder (approx. 0.7mg)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2[C@H](O3)[C@H](O)[C@H](OP(OC[C@@H]4[C@@H](O5)[C@@H](O)[C@H](N6C=NC7=C6N=CN=C7N)O4)(O)=O)[C@H]3COP5(O)=O.C

InChI

1S/C20H24N10O12P2/c21-15-9-17(25-3-23-15)29(5-27-9)19-11(31)13-7(39-19)1-37-43(33,34)42-14-8(2-38-44(35,36)41-13)40-20(12(14)32)30-6-28-10-16(22)24-4-26-18(10)30/h3-8,11-14,19-20,31-32H,1-2H2,(H,33,34)(H,35,36)(H2,21,23,25)(H2,22,24,26)/t7-,8-,11-,12-,13-,14-,19-,20-/m1/s1

InChI key

PDXMFTWFFKBFIN-XPWFQUROSA-N

Biochem/physiol Actions

It has a role in various signaling pathways. In Gram-positive bacteria, it influences cell growth and survival and modulates virulence. c-di-AMP is formed by diadenylate cyclases (DACs), which are cyclase domain-containing proteins.
c-di-AMP is a bacterial secondary messenger. c-di-AMP acts as a potent mucosal adjuvant stimulating both humoral and cellular responses.

Preparation Note

c-di-AMP is soluble in water (≥ 16 mM, limits have not been determined).

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qichao Sun et al.
Enzyme and microbial technology, 143, 109700-109700 (2020-12-31)
Cyclic di-AMP is a bacterial nucleotide second messenger and evaluated as a potential vaccine adjuvant candidate. Here, we report a practical and economical enzymatic method for gram-scale preparation of c-di-AMP using an immobilized Vibrio cholerae dinucleotide cyclase DncV. The method
c-di-AMP: An Essential Molecule in the Signaling Pathways that Regulate the Viability and Virulence of Gram-Positive Bacteria
Fahmi T, et al.
Genes, 8(8), 197-197 (2017)
Andrea Annibal et al.
Analytical and bioanalytical chemistry, 413(26), 6457-6468 (2021-09-04)
Cyclic dinucleotides (CDNs) are key secondary messenger molecules produced by cyclic dinucleotide synthases that trigger various cellular signaling cascades from bacteria to vertebrates. In mammals, cyclic GMP-AMP synthase (cGAS) has been shown to bind to intracellular DNA and catalyze the
W Michael Babinchak et al.
Nature communications, 11(1), 5574-5574 (2020-11-06)
Liquid-liquid phase separation (LLPS) of proteins that leads to formation of membrane-less organelles is critical to many biochemical processes in the cell. However, dysregulated LLPS can also facilitate aberrant phase transitions and lead to protein aggregation and disease. Accordingly, there

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service