Skip to Content
Merck
All Photos(1)

Key Documents

SMB01065

Sigma-Aldrich

Neolicuroside

≥85% (LC/MS-ELSD)

Synonym(s):

4-O-(beta-D-apiofuranosyl(1-2)-beta-D-glucopyranosyl)isoliquiritigenin, Isoliquiritigenin-4′-O-apiosyl(1→2)glucoside, Isoliquiritin apioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30O13
CAS Number:
Molecular Weight:
550.51
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

550.51

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

InChI

1S/C26H30O13/c27-10-19-20(32)21(33)22(39-25-23(34)26(35,11-28)12-36-25)24(38-19)37-15-5-1-13(2-6-15)3-8-17(30)16-7-4-14(29)9-18(16)31/h1-9,19-25,27-29,31-35H,10-12H2/b8-3+/t19-,20-,21+,22-,23+,24-,25+,26-/m1/s1

InChI key

VMMVZVPAYFZNBM-KVFWHIKKSA-N

General description

Neolicuroside, also known as Isoliquiritin apioside, is a chalcone glycoside structurally related to steroidal saponins. This bioactive plant metabolite is commonly derived from the roots of Glycyrrhiza Sp. plants and has demonstrated anti-inflammatory, anti-tussive, anti-tumor, antigenotoxic(1), anti-diabetic, and antioxidant properties.
Natural product derived from plant source.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Neolicuroside modulates signaling pathways (e.g., MAPK, NF-κB), impacting vital cell processes such as proliferation, differentiation, and apoptosis. Its multitargeted mechanism includes scavenging free radicals, suppressing pro-inflammatory cytokine production, inhibiting matrix metalloproteinase (MMP) activity and hindering cancer cell growth through apoptosis and cell cycle arrest. Furthermore, it also exerts immunomodulatory effects possibly by activating toll-like receptor 4 (TLR4).

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of antigenotoxic activity of isoliquiritin apioside from Glycyrrhiza glabra L
Kaur P, et al.
Toxicology in vitro, 23, 680-686 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service