Skip to Content
Merck
All Photos(1)

Key Documents

S2189

Sigma-Aldrich

SE 175

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO6S
CAS Number:
Molecular Weight:
347.34
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

shipped in

wet ice

storage temp.

−20°C

SMILES string

COC(=O)c1ccccc1SC(=O)c2ccc(CO[N+]([O-])=O)cc2

InChI

1S/C16H13NO6S/c1-22-15(18)13-4-2-3-5-14(13)24-16(19)12-8-6-11(7-9-12)10-23-17(20)21/h2-9H,10H2,1H3

InChI key

JDIVRBDMTYQVOK-UHFFFAOYSA-N

Application

SE 175 may be used in cell signaling studies.

Biochem/physiol Actions

SE 175 is a nitrate compound that belongs to the same class as nitroglycerine. It acts as a vasodilator and has clinical use in the treatment of angina. NO donors also confer neuroprotection and relieve neuropathic pain.
An organic nitrate compound that acts as an NO donor in vivo following reductive transformation of the nitrate group to nitric oxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Rybczynska et al.
Vaccine, 38(33), 5337-5342 (2020-06-24)
Freezing of alum-based vaccines drastically alters their colloidal composition and leads to irreversible cluster formation. The loss of stability is well described, but the impact of frost damage on the functionality of the induced and secreted antibody repertoire has not
Gregory R J Thatcher et al.
Free radical biology & medicine, 37(8), 1122-1143 (2004-09-29)
Nitroglycerine has been used clinically in the treatment of angina for 130 years, yet important details on the mechanism of action, biotransformation, and the associated phenomenon of nitrate tolerance remain unanswered. The biological activity of organic nitrates can be said

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service