Skip to Content
Merck
All Photos(1)

Key Documents

MHS80

Sigma-Aldrich

Mayer′s Hematoxylin Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

1 g/L

pH

2.4 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.

Application

General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.

Other Notes

1 g/L certified hematoxylin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Smith et al.
Molecular cancer therapeutics, 8(6), 1666-1675 (2009-06-11)
Although viridins like wortmannin (Wm) have long been examined as anticancer agents, their ability to self-activate has only recently been recognized. Here, we describe the cytostatic effects of a self-activating viridin (SAV), which is an inactive, polymeric prodrug. SAV self-activates
Nicola E Owen et al.
EBioMedicine, 56, 102794-102794 (2020-05-27)
BMP9, originating from the liver, and BMP10 are circulating BMPs that preserve vascular endothelial integrity. We assessed BMP9, BMP10 and soluble endoglin (sEng) levels and their relationships to liver disease severity and associated pulmonary vascular syndromes in a cohort of
J Jason Hoth et al.
Shock (Augusta, Ga.), 31(4), 376-381 (2008-07-31)
Blunt chest trauma resulting in pulmonary contusion with an accompanying acute inflammatory response is a common but poorly understood injury. We previously demonstrated that toll-like receptor 2 (TLR-2) participates in the inflammatory response to lung injury. We hypothesized that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service