Skip to Content
Merck
All Photos(1)

Key Documents

I7643

Millipore

Amine-terminated magnetic particles

50 mg/mL in deionized water

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

synthetic (organic)

form

suspension

concentration

50 mg/mL in deionized water

extent of labeling

~12 μmol per mL

technique(s)

affinity chromatography: suitable

matrix

Superparamagnetic iron oxide particles, approx. 1 μm in size

capacity

~10 mg/mL, protein coupling capacity (protein)

suitability

suitable for microbiology

storage temp.

2-8°C

Application

Amine-terminated magnetic particles are buffered in an aqueous solution and used for affinity chromatography, protein chromatography and activated/functionalized matrices. Amine-terminated magnetic particles have been used in the development of methods to detect Mycobacterium tuberculosis and Herpes Simplex Virus-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edith Torres-Chavolla et al.
Biosensors & bioelectronics, 26(11), 4614-4618 (2011-05-28)
The present study describes the development of a DNA based biosensor to detect Mycobacterium tuberculosis using thermophilic helicase-dependent isothermal amplification (tHDA) and dextrin coated gold nanoparticles (AuNPs) as electrochemical reporter. The biosensor is composed of gold nanoparticles (AuNPs) and amine-terminated
Feng Wang et al.
Bioresource technology, 101(23), 8931-8935 (2010-07-27)
Newly large-pore magnetic mesoporous silica nanoparticles (MMSNPs) with wormhole framework structures were synthesized for the first time by using tetraethyl orthosilicate as the silica source and amine-terminated Jeffamine surfactants as template. Iminodiacerate was attached on these MMSNPs through a silane-coupling
David A C Thomson et al.
The Analyst, 136(8), 1599-1607 (2011-03-04)
Amplification-free detection of nucleic acids in complex biological samples is an important technology for clinical diagnostics, especially in the case where the detection is quantitative and highly sensitive. Here we present the detection of a synthetic DNA sequence from Herpes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service