Skip to Content
Merck
All Photos(1)

Key Documents

76184

Sigma-Aldrich

3-Palmitoyl-sn-glycerol

≥99.0% (TLC)

Synonym(s):

MG(16:0/0:0/0:0), (R)-Glycerol 1-palmitate, L-α-Palmitin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O4
CAS Number:
Molecular Weight:
330.50
Beilstein:
1728236
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥99.0% (TLC)

form

solid

functional group

ester

lipid type

neutral glycerides

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](O)CO

InChI

1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,20-21H,2-17H2,1H3/t18-/m1/s1

InChI key

QHZLMUACJMDIAE-GOSISDBHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping-Yi Li et al.
Frontiers in microbiology, 11, 9-9 (2020-02-11)
Monoacylglycerol lipases (MGLs) are present in all domains of life. However, reports on bacterial MGLs are still limited. Until now, reported bacterial MGLs are all thermophilic/mesophilic enzymes from warm terrestrial environments or deep-sea hydrothermal vent, and none of them originates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service