Skip to Content
Merck
All Photos(1)

Key Documents

72678

Millipore

Bile esculin azide agar ISO 7899-2:2000

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to ISO 7899-2:2000

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bacteriological agar, 15.0 g/L
esculin, 1.0 g/L
ferric ammonium citrate, 0.5 g/L
ox bile, 10.0 g/L
peptone, 3.0 g/L
sodium azide, 0.15 g/L
sodium chloride, 5.0 g/L
tyrptone, 17.0 g/L
yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | specific enzyme detection: suitable

final pH

7.1±0.1 (25 °C)

application(s)

environmental
food and beverages
water monitoring

microbiology

suitability

Enterococcus spp.
Streptococcus spp.

Application

Selective medium for the isolation and presumptive identification of intestinal enterococci by Membrane filtration method.

Preparation Note

Suspend 56.6 grams of the medium in one liter of distilled water. Mix well and dissolve by heating with frequent agitation. Boil for one minute until complete dissolution. Dispense into appropriate containers and sterilize in autoclave at 121ºC for 15 minutes. Overheating can cause darkening of the medium. If tubes are used, allow cooling in a slanted position

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service